Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6H Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144099
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144099 Supplier Invitrogen™ Supplier No. PA5144099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LY6H Polyclonal Antibody for ICC/IF, IHC (P), ELISA

LY6H is highly expressed in particular subdivisions of human brain and also in MOLT-3 and -4 acute lymphoblastic leukemia cells. It has some isoforms with MW 14-16 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6H
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LY6H
Gene Accession No. O94772
Gene Alias LY6H; Ly-6H; lymphocyte antigen 6 complex, locus H; lymphocyte antigen 6H; NMLY6
Gene Symbols LY6H
Host Species Rabbit
Immunogen Recombinant Human Lymphocyte antigen 6H protein (26-115AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4062
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.