Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly6K Antibody (750747) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6648X
Description
Ly6K Monoclonal antibody specifically detects Ly6K in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Ly6K | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54742 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
750747 | |
Flow Cytometry, CyTOF-ready | |
cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
Chinese hamster ovary cell line CHO-derived recombinant human Ly6K, Met1-Gly138, Accession # Q17RY6 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction