Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572689
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572689 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572689 Supplier Invitrogen™ Supplier No. PA572689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LY6K Polyclonal Antibody for Western Blot, ELISA

Required for sperm migration into the oviduct and male fertility by controlling binding of sperm to zona pellucida. May play a role in cell growth (PubMed:18089789). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6K
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LY6K
Gene Accession No. Q17RY6, Q9CWP4
Gene Alias 2410015A16Rik; 3110035B01Rik; cancer/testis antigen 97; CO16; CT97; FLJ35226; HSJ001348; Ly6k; ly-6K; lymphocyte antigen 6 complex, locus K; lymphocyte antigen 6 complex, locus K-like; lymphocyte antigen 6K; mLy-6K; up-regulated in lung cancer 10; URLC10
Gene Symbols LY6K
Host Species Rabbit
Immunogen LY6K antibody for an 18 amino acid synthetic peptide near the amino terminus of human LY6K. The immunogen is within the first 50 amino acids of LY6K.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100363492, 54742, 76486
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.