Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529051
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes acid alpha-glucosidase, which is essential for the degradation of glycogen to glucose in lysosomes. Different forms of acid alpha-glucosidase are obtained by proteolytic processing. Defects in this gene are the cause of glycogen storage disease II, also known as Pompe's disease, which is an autosomal recessive disorder with a broad clinical spectrum. Three transcript variants encoding the same protein have been found for this gene.
Specifications
LYAG | |
Polyclonal | |
Unconjugated | |
Gaa | |
70 kDa lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; acid maltase; Aglucosidase alfa; E430018M07Rik; Gaa; glucosidase alpha, acid; glucosidase, alpha, acid; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); LYAG; Lysosomal alpha-glucosidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.76 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P10253 | |
Gaa | |
Recombinant fragment corresponding to a region within amino acids 683 and 952 of Human LYAG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction