Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544683 Supplier Thermo Scientific Supplier No. PA544683
Add to Cart
Add to Cart

LYAR Polyclonal Antibody

LYAR Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LYAR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LYAR
Gene Accession No. Q9NX58
Gene Alias apoptosis-related protein; cell growth-regulating nucleolar protein; hypothetical protein FLJ20425; Ly1 antibody reactive; Ly1 antibody reactive clone; Ly1 antibody reactive homolog; Ly1 antibody-reactive protein; Lyar; LYAR protein; MLZ-264; PNAS-5; protein expressed in male leptotene and zygotene spermatocytes 264; ZC2HC2; ZLYAR
Gene Symbols LYAR
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LYAR.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55646
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.