Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557444 Supplier Invitrogen™ Supplier No. PA557444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELLEQISAF DNVPRKKAKF QNWMKNSLKV HNESILDQVW NIFSEASNSE PVNKEQDQRP LHPVANPHAE ISTKVPASKV KDAVEQQGEV KKNKRERKEE R Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.

Plays a role in the maintenance of the appropriate processing of 47S/45S pre-rRNA to 32S/30S pre-rRNAs and their subsequent processing to produce 18S and 28S rRNAs (PubMed:24495227). Also acts at the level of transcription regulation. Along with PRMT5, binds the gamma-globin (HBG1/HBG2) promoter and represses its expression (PubMed:25092918). In neuroblastoma cells, may also repress the expression of oxidative stress genes, including CHAC1, HMOX1, SLC7A11, ULBP1 and SNORD41 that encodes a small nucleolar RNA (PubMed:28686580). Preferentially binds to a DNA motif containing 5'-GGTTAT-3' (PubMed:25092918). Negatively regulates the antiviral innate immune response by targeting IRF3 and impairing its DNA-binding activity (PubMed:31413131). In addition, inhibits NF-kappa-B-mediated expression of proinflammatory cytokines (PubMed:31413131). Stimulates phagocytosis of photoreceptor outer segments by retinal pigment epithelial cells. Prevents nucleolin/NCL self-cleavage, maintaining a normal steady-state level of NCL protein in undifferentiated embryonic stem cells (ESCs), which in turn is essential for ESC self-renewal. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYAR
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LYAR
Gene Accession No. Q9NX58
Gene Alias apoptosis-related protein; cell growth-regulating nucleolar protein; hypothetical protein FLJ20425; Ly1 antibody reactive; Ly1 antibody reactive clone; Ly1 antibody reactive homolog; Ly1 antibody-reactive protein; Lyar; LYAR protein; MLZ-264; PNAS-5; protein expressed in male leptotene and zygotene spermatocytes 264; ZC2HC2; ZLYAR
Gene Symbols LYAR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYAR. Recombinant protein control fragment (Product #RP-96673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.