Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lyb-2.1 Monoclonal Antibody (10.1.D2), APC
GREENER_CHOICE

Catalog No. PIMA528656
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528656 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528656 Supplier Invitrogen™ Supplier No. MA528656
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lyb-2.1 Monoclonal Antibody for Flow

CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyb-2.1
Applications Flow Cytometry
Classification Monoclonal
Clone 10.1.D2
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5
Gene Cd72
Gene Accession No. P21855
Gene Alias B-cell differentiation antigen CD72; CD72; CD72 antigen; CD72c; Ly-19; Ly32; Ly-32; Lyb-2; Ly-m19; lymphocyte antigen 32
Gene Symbols Cd72
Host Species Mouse
Immunogen DBA/2 mouse spleen cells.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12517
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.