Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lyb-2.1 Monoclonal Antibody (CT-72.1), Biotin
Mouse Monoclonal Antibody
$393.00 - $720.00
Specifications
Antigen | Lyb-2.1 |
---|---|
Clone | CT-72.1 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517925
|
Invitrogen™
MA517925 |
100 μg |
Each for $393.00
|
|
|||||
PIMA517926
|
Invitrogen™
MA517926 |
300 μg |
Each for $720.00
|
|
|||||
Description
Lyb-2.1 Monoclonal Antibody for Flow
CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.Specifications
Lyb-2.1 | |
0.1 mg/mL | |
Flow Cytometry | |
Biotin | |
Mouse | |
RUO | |
PBS with 0.02% sodium azide | |
P21855 | |
12517 | |
Mouse CD72.1. | |
Antibody |
CT-72.1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Mouse | |
Cd72 | |
B-cell differentiation antigen CD72; CD72; CD72 antigen; CD72c; Ly-19; Ly32; Ly-32; Lyb-2; Ly-m19; lymphocyte antigen 32 | |
Cd72 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title