Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 594

Catalog No. L32471
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32471 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32471 Supplier Invitrogen™ Supplier No. L32471
Add to Cart
Edge
Add to Cart

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 594, is a bright-red LEL-fluorophore conjugate.

The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 594 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

• Very bright DyLight 594 conjugate detected with traditional Texas Red filter set, Ex/Em: 592/617 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Tomato
Conjugate DyLight 594
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.

Label Type DyLight Fluors
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins
Product Line DyLight

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.