Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558685
Description
Immunogen sequence: GSYPFSHSMK PHLHPRLYHG CYGDIMTMKT SGATCDANSV MNCGIRGSEM FAEMDLRAIK PYQTLIKEVG QRHCVDPA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.

Specifications
LYG2 | |
Polyclonal | |
Unconjugated | |
LYG2 | |
LYG2; LYGH; LYSG2; lysozyme g2; lysozyme G-like 2; lysozyme g-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SG7 | |
LYG2 | |
Recombinant protein corresponding to Human LYG2. Recombinant protein control fragment (Product #RP-96217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction