Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112336
Description
Specificity: Human LYG2.
Specifications
| LYG2 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q86SG7 | |
| LYG2 | |
| E. coli-derived Human LYG2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LYG2 | |
| LYG2; LYGH; LYSG2; lysozyme g2; lysozyme G-like 2; lysozyme g-like protein 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 254773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction