Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYG2 Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305997AF405
Description
LYG2 Polyclonal specifically detects LYG2 in Human samples. It is validated for Western Blot.Specifications
LYG2 | |
Polyclonal | |
Western Blot | |
EC 3.2.1.-, LYGHMGC119046, lysozyme G-like 2, lysozyme g-like protein 2, MGC119047, MGC119049 | |
Produced in rabbits immunized with E. coli-derived Human LYG2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
254773 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction