Learn More
Invitrogen™ LYK5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587821
Description
Immunogen sequence: FLPEGGCYEL LTVIGKGFED LMTVNLARYK PTGEYVTVRR INLEACSNEM VTFLQGELHV SKLFNHPNIV PYRATFIADN ELWVVTSFMA YGSAKDLICT HFMDGMNELA IAYILQGVLK ALDYIHHMGY VHRSVKASHI LISVDGKVYL SGLRSNLSMI SHGQRQRVVH DFPKYSVKVL PWLSPE; Positive Samples: Mouse skeletal muscle; Cellular Location: Cytoplasm, Nucleus.
LYK5; STE20-related Adaptor Protein (STRAD); FLJ90524. Gene Map Locus is 17q23.3 and members of STE-20 like kinase family are known to stimulate MAPK pathways by directly activating MAPKKK. STRAD is a novel pseudokinase member of this family consisting of a STE-20 like kinase domain but lacks several residues that are required for its catalytic activity. It specifically binds LKB1 and plays a key role in regulating tumor suppressor activities of LKB1. It functions as an upstream activator of LKB1 and also directs the sub-cellular localization of LKB1 by anchoring it in the cytoplasm. STRAD-LKB1 interaction results in phosphorylation of STRAD and enhanced autophosphorylation of LKB1.
Specifications
LYK5 | |
Polyclonal | |
Unconjugated | |
STRADA | |
2610019A05Rik; 6030402H20Rik; AI480680; E130112C09Rik; LYK5; NY-BR-96; PMSE; protein kinase LYK5; serologically defined breast cancer antigen NY-BR-96; STE20-like pseudokinase; STE20-related adapter protein; STE20-related kinase adapter protein alpha; STE20-related kinase adaptor alpha; Stlk; Strad; STRAD alpha; Strada | |
Rabbit | |
Affinity chromatography | |
RUO | |
72149 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.98 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UUJ4 | |
STRADA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-210 of human STRADA (NP_6991662). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.