Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphatic Endothelial Cells Antibody (LA102) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018060 0.02 mg
NB11096416 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018060 Supplier Novus Biologicals Supplier No. NB110964160.02MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Lymphatic Endothelial Cells Monoclonal specifically detects Lymphatic Endothelial Cells in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Lymphatic Endothelial Cells
Applications Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone LA102
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.2) with No Preservative
Gene Alias LEC26
Host Species Rat
Immunogen with collagenase- and neuraminidase-treated mouse benign lymphangiomas
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity LA102 reacts with mouse lymphatic vessels except the thoracic duct and the marginal sinus of lymph nodes, but not with any blood vessels. LA102 recognized a protein of around 26 kDa.The antigen recognized by LA102 was localized on both luminal and abluminal lymphatic endothelial cell membranes. Besides lymphatics, some lymphoid cells are also recognized.
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.