Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lymphocytes/Langerhans Cells Monoclonal Antibody (MsGp2)
GREENER_CHOICE

Catalog No. PIMA528470
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528470 Supplier Invitrogen™ Supplier No. MA528470
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an antigen present on the majority of T and B lymphocytes and thymocytes, although the antigen is not expressed by germinal center cells of the lymph node.

Lymphocytes/Langerhans Cells antibody recognizes an antigen present on the majority of T and B lymphocytes and thymocytes, although the antigen is not expressed by germinal center cells of the lymph node.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lymphocytes/Langerhans Cells
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone MsGp2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Nylon wool adherent B cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.