Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lymphotactin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563473 Supplier Invitrogen™ Supplier No. PA563473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VCADPQATWV RDVVRSMDRK SNTRNNMIQT KPTGTQQSTN TAVTLTG Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 40%.

Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lymphotactin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Xcl1
Gene Accession No. P47992
Gene Alias AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM1alpha; SCM-1alpha; SCM-1-alpha; SCM1beta; SCM-1beta; SCM-1-beta; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1
Gene Symbols Xcl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Lymphotactin. Recombinant protein control fragment (Product #RP-104515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6375
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.