Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Lymphotoxin-alpha/TNF-beta Antibody (3G11), Novus Biologicals™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | Lymphotoxin-alpha/TNF-beta | 
|---|---|
| Clone | 3G11 | 
| Dilution | ELISA | 
| Applications | ELISA | 
| Classification | Monoclonal | 
Description
Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for ELISASpecifications
| Lymphotoxin-alpha/TNF-beta | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| A synthesized peptide derived from Human Lymphotoxin-alpha/TNF-beta [UniProt P01374] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | 
| 3G11 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 4049 | |
| IgG | |
| Affinity purified | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            