Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin-alpha/TNF-beta Antibody (SR1300), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321751100UL
Description
Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Western BlotSpecifications
Lymphotoxin-alpha/TNF-beta | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1300 | |
Western Blot 1:500-1:2000 | |
LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
A synthesized peptide derived from human Lymphotoxin-alpha/TNF-beta (Uniprot #: P01374) | |
100 μg | |
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
4049 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction