Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (4H8 WH2) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280054
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Flow Cytometry, Block/Neutralize.Specifications
| Lymphotoxin beta R/TNFRSF3 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Functional | |
| CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes mouse Lymphotoxin beta R/TNFRSF3. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Flow Cytometry, Functional Assay | |
| 4H8 WH2 | |
| Unconjugated | |
| PBS containing 10% glycerol | |
| LTBR | |
| Recombinant mouse Lymphotoxin beta R/TNFRSF3 (cysteine-rich region/aa 31-221). | |
| 100 μg | |
| Apoptosis, Cytokine Research | |
| 4055 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction