Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (71319), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB629M
This item is not returnable.
View return policy
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal antibody specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
Lymphotoxin beta R/TNFRSF3 | |
Monoclonal | |
DyLight 594 | |
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
Mouse myeloma cell line NS0-derived recombinant human Lymphotoxin beta R/TNFRSF3, Gln31-Met227, Accession # P36941 | |
0.5 mL | |
Apoptosis, Cytokine Research | |
4055 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
71319 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction