Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (MM0461-3T24) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Lymphotoxin beta R/TNFRSF3 |
---|---|
Clone | MM0461-3T24 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Lymphotoxin beta R/TNFRSF3 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis, Cytokine Research | |
P36941 | |
4055 | |
IgG1 | |
Protein G purified |
MM0461-3T24 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
LTBR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title