Learn More
Promega LYN B Kinase Enzyme System, 1mg
Catalog No. PRV3712 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3712 1 mg
1 options
Catalog No. PRV3712 Supplier Promega Supplier No. V3712
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human LYN B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LYN is a 56kDa tyrosine kinase that is similar to mouse T-lymphocyte-specific tyrosine kinase p56lck, v-yes, v-fgr and v-src.
  • Easily Screen and Profile LYN B Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C