Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lyn Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529329 Supplier Invitrogen™ Supplier No. PA529329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse brain, K562. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (85%), Xenopus laevis (90%), Chicken (92%), Sheep (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyn
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene LYN
Gene Accession No. P07948, P25911
Gene Alias AA407514; CD180; CD180 antigen; CD180 molecule; CH73-38P6.3; FLJ26625; Hck-2; JTK8; lck/Yes-related novel protein tyrosine kinase; LY64; Ly78; lymphocyte antigen 64; lymphocyte antigen-64, radioprotective, 105kDa; LYN; lyn protein non-receptor kinase; lyn protein tyrosine kinase; LYN proto-oncogene, Src family tyrosine kinase; p53Lyn; p56Lyn; Radioprotective 105 kDa protein; RP105; Tyrosine-protein kinase Lyn; v-yes-1 Yamaguchi sarcoma viral related oncogene homolog; v-yes-1 Yamaguchi sarcoma viral related oncogene protein-like protein; Yamaguchi sarcoma viral (v-yes-1) oncogene homolog; zgc:92124
Gene Symbols LYN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LYN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17096, 4067
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.