Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyn Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985000
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5000 Supplier Bioss Supplier No. BS2906R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).

Specifications

Antigen Lyn
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LYN
Gene Accession No. P07948
Gene Alias AA407514; BOS_13851; CH73-38P6.3; FLJ26625; Hck-2; JTK8; Lck/Yes-related novel protein tyrosine kinase; LOC100340056; LYN; lyn protein non-receptor kinase; LYN proto-oncogene, Src family tyrosine kinase; p53Lyn; p56Lyn; Tyrosine-protein kinase Lyn; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog; v-yes-1 Yamaguchi sarcoma viral related oncogene protein-like protein; Yamaguchi sarcoma viral (v-yes-1) oncogene homolog; zgc:92124
Gene Symbols LYN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Lyn.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4067
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.