Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYN Rabbit anti-Human, Unconjugated, Clone: IFI-12, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20536
Description
LYN Monoclonal antibody specifically detects LYN in Human samples. It is validated for ICC, WB-Ce, IHC-P, Flow Cyt.Specifications
LYN | |
Monoclonal | |
Unconjugated | |
LYN | |
Rabbit | |
100 μg | |
4067 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
IFI-12 | |
v-yes-1 Yamaguchi sarcoma viral related oncogene homolog | |
FLJ26625, JTK8 | |
A synthetic phosphopeptide corresponding to residues surrounding Y396 of human LYN | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction