Learn More
LYNX1 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111850

Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: CAYNGDN(S) FKPMR Homology: Mouse, rat - 12/13 amino acid residues identical; human - 11/13 amino acid residues identical.
LYNX1 (extracellular) Polyclonal antibody specifically detects LYNX1 (extracellular) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Specifications
LYNX1 (extracellular) | |
Polyclonal | |
Unconjugated | |
Lynx1 | |
AI838844; Endogenous prototoxin LYNX1; GC26; Ly-6 neurotoxin-like protein 1; Ly6/neurotoxin 1; ly-6/neurotoxin-like protein 1; Lynx1; secreted Ly-6/uPAR domain-containing protein 2; secreted Ly6/uPAR related protein 2; secreted Ly-6/uPAR-related protein 2; SLURP2; SLURP-2; Testicular tissue protein Li 112 | |
Peptide corresponding to amino acid residues 26-38 of mouse Lynx1 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P0DP58, P0DP60 | |
Rabbit | |
Affinity Purified | |
RUO | |
23936, 300018, 66004 | |
-20°C | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.