Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lyo-ready Bst DNA Polymerase

Catalog No. p-7229401
Encompass_Preferred
Click to view available options
Quantity:
1200 U
6000 U
Concentration:
40 U/μL
6 U/μL
This item is not returnable. View return policy
This item is not returnable. View return policy

Invitrogen Lyo-ready Bst DNA Polymerase is a proprietary mutant of Bst DNA polymerase, large fragment, with a fast reaction speed, increased sensitivity, and increased tolerance to inhibitors.

Invitrogen Lyo-ready Bst DNA Polymerase is a proprietary mutant of Bst DNA polymerase, large fragment, with a fast reaction speed, increased sensitivity, and increased tolerance to inhibitors. By retaining strong strand-displacement activity, this enzyme helps enable highly efficient isothermal amplifications such as loop-mediated isothermal amplification (LAMP).

Invitrogen Lyo-ready Bst DNA Polymerase is a proprietary mutant of Bst DNA polymerase, large fragment, with a fast reaction speed, increased sensitivity, and increased tolerance to inhibitors. By retaining strong strand-displacement activity, this enzyme helps enable highly efficient isothermal amplifications such as loop-mediated isothermal amplification (LAMP).

Lyo-ready Bst DNA Polymerase possesses 5´→3´ DNA polymerase activity and lacks 5´→3´ exonuclease activity and 3´→5´ exonuclease activity. It is also a glycerol-free enzyme, enabling assay lyophilization and use in microfluidics-based technologies.

Lyo-ready Bst DNA Polymerase features
• Fast target detection in as little as 10 minutes
• Enhanced sensitivity down to 50 copies using optimized reaction buffer in (RT-)LAMP reaction
• Robust amplification, even from inhibitor-containing RNA or DNA samples
• Ability to optimize LAMP or RT-LAMP reactions to fit your needs

Applications
Lyo-ready Bst DNA Polymerase can be used for a wide variety of applications including:
• Loop-mediated isothermal amplification (LAMP)
• Whole genome amplification (WGA)
• Rolling circle amplification (RCA)
• Recombinase polymerase amplification (RPA)
• Helicase-dependent amplification (HDA)
• Exponential amplification reaction (EXPAR)

Notes
1. If performing real-time LAMP detection with SYTO 9 Green Fluorescent Nucleic Acid Stain, dilute the stock to 50 μM with nuclease-free water before every use.
2. SuperScript IV Reverse Transcriptase and RNaseOUT Recombinant Ribonuclease Inhibitor are recommended for reactions containing RNA targets.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stand-alone enzyme
For Use With (Application) LAMP, RT-LAMP
Polymerase Lyo-ready Bst DNA Polymerase
Quantity 1200 U
Content And Storage

• Lyo-ready Bst DNA Polymerase (200 μL at 6 U/μL)
• 10X Bst Reaction Buffer (1 mL)
• 200 mM MgCl2 (1 mL)
• 5 M Betaine (1 mL)

Store at –15 to –25°C.

No. of Reactions 200 Reactions
Optimal Reaction Temperature 65°C
Shipping Condition Dry Ice
Concentration 6 U/μL
Reaction Time ≤10 minutes
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.