Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA)

Catalog No. p-200087434
Encompass_Preferred
Click to view available options
Quantity:
100 Reactions
120 μg
1600 μg
2000 μL
500 Reactions
Product Type:
Lyo-ready RPA Kit
Lyo-ready T4 Gene 32 Protein
Lyo-ready T4 UvsX Protein
Lyo-ready T4 UvsY Protein
Reaction Buffer

Invitrogen Lyo-ready RPA reagents are designed for fast, sensitive, and specific DNA amplification through isothermal recombinase polymerase amplification (RPA) reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats.

Invitrogen Lyo-ready recombinase polymerase amplification (RPA) reagents are designed for fast, sensitive, and specific DNA amplification through isothermal RPA reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats. Reverse transcriptase can be added to the RPA reaction for amplification of RNA targets. All reagents are glycerol-free, which enables assay lyophilization.

Features of Lyo-ready RPA products

  • Enhanced sensitivity down to 1 copy using optimized (RT-)RPA reaction buffer
  • Robust and specific RNA/DNA amplification
  • Amplification from inhibitor-containing samples
  • Fast target amplification in as little as 20 minutes
  • Ability to easily optimize RPA or RT-RPA reactions

Kit components

The Lyo-ready RPA Kit includes all required components for an RPA reaction:

  • Lyo-ready T4 UvsX Protein—produced from E. coli cells bearing cloned uvsX gene derived from bacteriophage T4
  • Lyo-ready T4 UvsY Protein—a recombination mediator protein that stimulates homologous recombination performed by the UvsX recombinase
  • Lyo-ready Bst DNA Polymerase (Cat. No. A56655)—a proprietary mutant of Bst DNA Polymerase, Large Fragment, with fast reaction speed, increased sensitivity, and tolerance to inhibitors
  • Lyo-ready T4 Gene 32 Protein—a single-stranded DNA binding protein produced from E. coli cells carrying cloned bacteriophage T4 gene 32
  • Optimized reaction buffer, magnesium chloride, and dNTPs
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lyo-ready T4 Gene 32 Protein
Form Liquid
For Use With (Application) RPA, RT-RPA, cDNA Synthesis, PCR
Quantity 1600 μg
Research Category Nucleic Acid Amplification
Content And Storage Lyo-ready T4 Gene 32 Protein, 80 μL, Store at -25°C to -15°C.
Shipping Condition Dry Ice
Concentration 20 mg/mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.