Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyp/PTPN22 Antibody, Novus Biologicals™
SDP

Catalog No. p-200040863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433903 25ul
NBP181276 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433903 Supplier Novus Biologicals Supplier No. NBP18127625UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lyp/PTPN22 Polyclonal specifically detects Lyp/PTPN22 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Lyp/PTPN22
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 3.1.3.48, Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP, Lymphoid phosphatase, lymphoid-specific protein tyrosine phosphatase, LYP, Lyp1, LYP2, PEP, PEST-domain phosphatase, protein tyrosine phosphatase, non-receptor type 22 (lymphoid), protein tyrosine phosphatase, non-receptor type 8, PTPN8LYP1, tyrosine-protein phosphatase non-receptor type 22
Gene Symbols PTPN22
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:SLLNQESAVLATAPRIDDEIPPPLPVRTPESFIVVEEAGEFSPNVPKSLSSAVKVKIGTSLEWGGTSEPKKFDDSVILRPSKSVKLRSPKSELHQDRSSPPPPLPERTLESFFLADEDCMQ
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 26191
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.