Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597943
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597943 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597943 Supplier Invitrogen™ Supplier No. PA597943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYPD3 Polyclonal Antibody for Western Blot, ELISA

Lymphocyte antigen 6 family member D3 (LYPD3), also known as C4.4A, is a gene located on chromosome 19q13.11 that encodes a glycosylphosphatidylinositol (GPI)-anchored protein belonging to the Ly6/uPAR superfamily. LYPD3 is expressed in various epithelial tissues and is implicated in cellular adhesion and migration processes. The protein plays a significant role in tumor progression and metastasis, with its expression being notably upregulated in several cancers, including squamous cell carcinoma and lung cancer. LYPD3 contributes to the invasive behavior of cancer cells by interacting with matrix components and modulating signaling pathways related to cell motility and adhesion. Its involvement in cancer biology makes LYPD3 a potential biomarker for tumor aggressiveness and a target for therapeutic intervention aimed at inhibiting cancer cell dissemination. Research continues to explore its functional mechanisms in oncogenesis and potential applications in cancer treatment strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LYPD3
Gene Accession No. O95274, Q91YK8
Gene Alias 2310061G07Rik; C4.4A; GPI-anchored metastasis-associated protein C4.4A homolog; GPI-anchored metastasis-associated protein homolog; LY6/PLAUR domain containing 3; ly6/PLAUR domain-containing protein 3; LYPD3; Matrigel-induced gene C4 protein; MIG-C4; UNQ491/PRO1007
Gene Symbols LYPD3
Host Species Rabbit
Immunogen Recombinant Human Ly6/PLAUR domain-containing protein 3 protein (31-326AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27076, 72434
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.