Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114140 Supplier Invitrogen™ Supplier No. PA5114140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGPQHLRLVQ LFCLLGAIST LPRAGALLCY EATASRFRAV AFHNWKWLLM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 79%; Rat: 79%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LYPD4
Gene Accession No. Q6UWN0
Gene Alias 4933400F01Rik; LY6/PLAUR domain containing 4; ly6/PLAUR domain-containing protein 4; LYPD4; SMR; sperm membrane receptor; testicular tissue protein Li 159; UNQ2552/PRO6181
Gene Symbols LYPD4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LYPD4 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147719
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.