Learn More
Invitrogen™ LYPD4 Polyclonal Antibody

Description
LYPD4 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | LYPD4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LYPD4 |
| Gene Accession No. | Q6UWN0, Q8BVP6 |
| Gene Alias | 4933400F01Rik; LY6/PLAUR domain containing 4; ly6/PLAUR domain-containing protein 4; LYPD4; SMR; sperm membrane receptor; testicular tissue protein Li 159; UNQ2552/PRO6181 |
| Gene Symbols | LYPD4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.