Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYPD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563001
Description
Immunogen sequence: QCYSFEHTYF GPFDLRAMKL PSISCPHECF EAILSLDTGY RAPVTLVRKG CWTGPPAGQT QSNPDALPPD YSVVRGCTTD KCNAHLMTHD ALPNLSQAP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.
The specific function of this protein remains unknown.
Specifications
LYPD5 | |
Polyclonal | |
Unconjugated | |
LYPD5 | |
LY6/PLAUR domain containing 5; ly6/PLAUR domain-containing protein 5; LYPD5; metastasis-associated protein; PRO4356; UNQ1908/PRO4356 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWN5 | |
LYPD5 | |
Recombinant protein corresponding to Human LYPD5. Recombinant protein control fragment (Product #RP-103768). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction