Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544431 Supplier Invitrogen™ Supplier No. PA544431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WTGPPAGQTQ SNADALPPDY SVVRGCTTDK CNAHLMTHDA LPNLSQAPDP Sequence homology: Human: 100%.

The specific function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPD5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LYPD5
Gene Accession No. Q6UWN5
Gene Alias LY6/PLAUR domain containing 5; ly6/PLAUR domain-containing protein 5; LYPD5; metastasis-associated protein; PRO4356; UNQ1908/PRO4356
Gene Symbols LYPD5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LYPD5 (aa 78-127).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284348
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.