Learn More
Invitrogen™ LYPLA1 Polyclonal Antibody

Description
Recommended positive controls: LYPLA1 tansfected 293T cell. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (80%), Rabbit (97%), Sheep (94%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | LYPLA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | LYPLA1 |
| Gene Accession No. | O75608 |
| Gene Alias | 25KDL; Acyl-protein thioesterase 1; APT1; APT-1; CaIPLA2; calcium-independent phospholipase A2; calcium-independent phospholipase A2 isoform 1; calcium-independent phospholipase A2 isoform 2; hAPT1; hypothetical protein LOC550279; LPL1; LPL-I; Lypla1; lysophopholipase 1; lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; LYSOPLA; lysoPLA I; phospholipase 1a; Pla1a; zgc:110260 |
| Gene Symbols | LYPLA1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.