Learn More
Invitrogen™ LYPLA1 Polyclonal Antibody

Description
Immunogen sequence: LTTQQKLAGV TALNCWLPLW ASFPQGPIGG ANRDISILQC HGDCDPLV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifications
Specifications
| Antigen | LYPLA1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LYPLA1 |
| Gene Accession No. | O75608 |
| Gene Alias | 25KDL; Acyl-protein thioesterase 1; APT1; APT-1; CaIPLA2; calcium-independent phospholipase A2; calcium-independent phospholipase A2 isoform 1; calcium-independent phospholipase A2 isoform 2; hAPT1; hypothetical protein LOC550279; LPL1; LPL-I; Lypla1; lysophopholipase 1; lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; LYSOPLA; lysoPLA I; phospholipase 1a; Pla1a; zgc:110260 |
| Gene Symbols | LYPLA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.