Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPLA1 Recombinant Rabbit Monoclonal Antibody (JE47-60)
GREENER_CHOICE

Catalog No. PIMA534855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534855 Supplier Invitrogen™ Supplier No. MA534855
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse stomach tissue, F9, rat kidney tissue, human prostate cancer tissue, human placenta tissue, mouse small intestine tissue.

Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine in both monomeric and micellar forms. The use of alternate polyadenylation sites has been found for this gene. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPLA1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE47-60
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene LYPLA1
Gene Accession No. O75608
Gene Alias 25KDL; Acyl-protein thioesterase 1; APT1; APT-1; CaIPLA2; calcium-independent phospholipase A2; calcium-independent phospholipase A2 isoform 1; calcium-independent phospholipase A2 isoform 2; hAPT1; hypothetical protein LOC550279; LPL1; LPL-I; Lypla1; lysophopholipase 1; lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; LYSOPLA; lysoPLA I; phospholipase 1a; Pla1a; zgc:110260
Gene Symbols LYPLA1
Host Species Rabbit
Immunogen Recombinant protein within Human Lysophospholipase 1 aa 1-160.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10434
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.