Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPLA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590709 Supplier Invitrogen™ Supplier No. PA590709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWLPLHRAFP QAANGSAKDL AILQCHGELD PMVPVRFGAL TAEKLRSVVT PARVQFKTYP GVMHSSCPQE MAAVKEFLEK LLPPV; Positive Samples: 293T, HT-29, Raji, Mouse thymus, Mouse brain, Rat lung; Cellular Location: Cytoplasm.

Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPLA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LYPLA2
Gene Accession No. O95372, Q9QYL8, Q9WTL7
Gene Alias Acyl-protein thioesterase 2; APT2; APT-2; DJ886K2.4; LPL-II; Lypla2; LysoII; Lysophospholipase 2; lysophospholipase II; lysoPLA II; mLyso II; testicular tissue protein Li 3
Gene Symbols LYPLA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 147-231 of human LYPLA2 (NP_0091911).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11313, 26394, 83510
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.