Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPLA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559682 Supplier Invitrogen™ Supplier No. PA559682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVAKTLRVLA SGDNNRIPVI GPLKIREQQR SAVSTSWLLP YNYTWSPEKV FVQTPTINYT LRDYRKFFQD IGFED Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine to glycerophosphorylcholine and a free fatty acid. This enzyme is present in the plasma and thought to be associated with high-density lipoprotein. A later paper contradicts the function of this gene. It demonstrates that this gene encodes a lysosomal enzyme instead of a lysophospholipase and has both calcium-independent phospholipase A2 and transacylase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPLA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G15
Gene Accession No. Q8NCC3
Gene Alias 1-O-acylceramide synthase; 1-O-acylceramide synthase {ECO:0000303; 1-O-acylceramidesynthase; ACS; ACS {ECO:0000303; C87498; EMBL:AAH98894.1}; EMBL:AAS66767.1}; Ensembl:ENSRNOP00000026996, ECO:0000312; Group XV phospholipase A2; group XV phospholipase A2 {ECO:0000312; GXVPLA2; LCAT-like lysophospholipase; LCAT-like lysophospholipase {ECO:0000250; LLPL; LLPL {ECO:0000250; LPLA2; LPLA2 {ECO:0000303; LYPLA3; Lysophospholipase 3; lysophospholipase 3 (lysosomal phospholipase A2); lysophospholipase 3 {ECO:0000250; lysosomal phospholipase A2; lysosomal phospholipase A2 {ECO:0000303; phospholipase A2 group XV; phospholipase A2, group XV; PLA2G15; pla2g15 {ECO:0000312; PubMed:15294901, ECO:0000312; PubMed:15294901}; RGD:1302982}; UniProtKB:Q8VEB4}; UNQ341/PRO540
Gene Symbols PLA2G15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYPLA3. Recombinant protein control fragment (Product #RP-98629).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23659
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.