Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYPLA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA531701
Description
Recommended positive controls: 293T, A431, Jurkat. Predicted reactivity: Mouse (92%), Rat (91%), Dog (96%), Rhesus Monkey (100%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine to glycerophosphorylcholine and a free fatty acid. This enzyme is present in the plasma and thought to be associated with high-density lipoprotein. A later paper contradicts the function of this gene. It demonstrates that this gene encodes a lysosomal enzyme instead of a lysophospholipase and has both calcium-independent phospholipase A2 and transacylase activities.
Specifications
| LYPLA3 | |
| Polyclonal | |
| Unconjugated | |
| PLA2G15 | |
| 1-O-acylceramide synthase; 1-O-acylceramide synthase {ECO:0000303; 1-O-acylceramidesynthase; ACS; ACS {ECO:0000303; C87498; EMBL:AAH98894.1}; EMBL:AAS66767.1}; Ensembl:ENSRNOP00000026996, ECO:0000312; Group XV phospholipase A2; group XV phospholipase A2 {ECO:0000312; GXVPLA2; LCAT-like lysophospholipase; LCAT-like lysophospholipase {ECO:0000250; LLPL; LLPL {ECO:0000250; LPLA2; LPLA2 {ECO:0000303; LYPLA3; Lysophospholipase 3; lysophospholipase 3 (lysosomal phospholipase A2); lysophospholipase 3 {ECO:0000250; lysosomal phospholipase A2; lysosomal phospholipase A2 {ECO:0000303; phospholipase A2 group XV; phospholipase A2, group XV; PLA2G15; pla2g15 {ECO:0000312; PubMed:15294901, ECO:0000312; PubMed:15294901}; RGD:1302982}; UniProtKB:Q8VEB4}; UNQ341/PRO540 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.64 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NCC3 | |
| PLA2G15 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 217 of Human LYPLA3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction