Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPLAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561097 Supplier Invitrogen™ Supplier No. PA561097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMKGGISNVW FDRFKITNDC PEHLESIDVM CQVLTDLIDE EVKSGIKKNR ILIGGFSMGG CMAMHLAYRN HQDVAGVFAL SSF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

LYPLAL1 does not exhibit phospholipase nor triacylglycerol lipase activity. It is able to hydrolyze only short chain substrates due to its shallow active site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPLAL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lyplal1
Gene Accession No. Q5VWZ2
Gene Alias BC027340; KIAA1238; LYPLAL1; lysophospholipase like 1; lysophospholipase-like 1; lysophospholipase-like protein 1; Q96AV0; Q96AVO
Gene Symbols Lyplal1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYPLAL1. Recombinant protein control fragment (Product #RP-94278).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.