Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra Parainfluenza Molecular Assay
SDP

Catalog No. 23044009 Shop All QuidelOrtho Products
Click to view available options
Quantity:
96 Tests/Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Lyra parainfluenza virus assay is a multiplex RT-PCR assay for qualitative detection/identification of viral nucleic acids extracted from nasal/nasopharyngeal swabs from symptomatic patients, Shown to detect and differentiate parainfluenza virus types 1, 2 and 3

  • RT-PCR
  • One-step reagent setup
  • Results in <75 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Long shelf-life
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls
  • Ability to customize panel of results

Specifications

Certifications/Compliance FDA approved*
Content And Storage 2°C to 8°C
Control Sets Included
CE Marker Yes
Clia Complexity High
DoA Calibrators No
Sample Type Nasal or Nasopharyngeal Swab
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) FDA approved for use on Applied Biosystems™ 7500 Fast Dx Real-Time PCR Instrument, QuantStudioTM Dx Real-Time PCR Instrument and Cepheid™ SmartCycler™ II
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.