Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Lyra Direct SARS COV-2 Assay
SDP

Catalog No. 23044173 Shop All QuidelOrtho Products
Change view
Click to view available options
Includes:
Control Assay
Viral Transport Media, Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-044-173 Viral Transport Media, Tubes
23-044-174 Control Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-173 Supplier Quidel Supplier No. M940
Add to Cart
Add to Cart
This item is not returnable. View return policy

A real-time RT-PCR assay intended for the in vitro qualitative detection of human coronavirus SARS-CoV-2 from viral RNA extracted from nasal, nasopharyngeal (NP) or oropharyngeal (OP) swab specimens from patients with signs and symptoms of COVID-19.

  • The Assay targets the non-structural Polyprotein (pp1ab) of the SARS-CoV-2 virus
  • One-step reagent set-up
  • Results in less than 70 minutes after extraction
  • Simplified and uniform workflow with standard pipetting volumes
  • No freezer needed. 2°C to 8°C storage

Specifications

CE Marker Yes
Clia Complexity High
Detectable Analytes SARS-CoV-2
DoA Calibrators No
Format 96-well Plate
Disposable Single-use
For Use With (Application) RT-PCR
Includes Viral Transport Media, Tubes
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.