Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel™ Lyra Direct SARS-CoV-2 Molecular Assay
Shop All QuidelOrtho Products

Click to view available options
Includes:
Nasal/oropharyngeal swabs, transport tubes, deep well plates
Nasal/oropharyngeal swabs, transport tubes, microfuge tubes
Nasopharyngeal swabs, transport tubes, deep well plates
Nasopharyngeal swabs, transport tubes, microfuge tubes
Tests and Controls
Sample Type:
Nasal, Nasopharyngeal, Oropharyngeal Swab
Nasal/oropharyngeal Swab
Nasopharyngeal Swab
Description
- RT-PCR
- One-step reagent setup
- Results in <70 minutes after extraction
- No freezer necessary
- Refrigration storage
Specifications
Specifications
Certifications/Compliance | Emergency Use Authorization (EUA) |
Content And Storage | 2°C to 8°C |
Control Sets | Included |
CE Marker | Yes |
Clia Complexity | High |
Detectable Analytes | SARS-CoV-2 |
DoA Calibrators | No |
Sample Type | Nasal, Nasopharyngeal, Oropharyngeal Swab |
Sensitivity | Nasopharyngeal: PPA – 29/30 = 97% (95% CI: 83.3 to 99.4%), Oropharyngeal: PPA – 15/15 = 100% (95% CI: 79.6 to 100%) |
Type | SARS-CoV-2 Assay |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction