Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra Influenza A+B Molecular Assay
SDP

Catalog No. 23044000 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
96 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-000 96 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-000 Supplier Quidel™ Supplier No. M100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lyra influenza A+B assay real-time PCR for qualitative detection/ID of viral nucleic acids extracted from nasal/nasopharyngeal swab specimens, FDA approved for use on Applied Biosystems™ 7500 FastDx RT-PCR, QuantStudioDx RT-PCR, and Cepheid™ SmartCycler™ II instruments

  • RT-PCR
  • One-step reagent setup
  • Results in <75 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Long shelf-life
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls
  • Ability to customize panel of results

Specifications

Certifications/Compliance FDA approved*
Content And Storage 2°C to 8°C
Control Sets Included
CE Marker Yes
Clia Complexity High
Detectable Analytes Influenza A/B viral RNA
Sample Type Nasal or Nasopharyngeal Swab
Sensitivity A: 100% B: 98.4%
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) FDA approved for use on Applied Biosystems™ 7500 Fast Dx Real-Time PCR Instrument, QuantStudioTM Dx Real-Time PCR Instrument and Cepheid™ SmartCycler™ II
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Specificity A: 98.7%, B: 95.5%
Test Time <75 min. after extraction
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.