Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra RSV+hMPV Molecular Assay
SDP

Catalog No. 23044002 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
96 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-002 96 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-002 Supplier Quidel™ Supplier No. M103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multiplex RT-PCR assay for qualitative detection/ID of respiratory syncytial virus/human metapneumovirus viral RNA extracted from nasal/nasopharyngeal swab specimens, FDA approved for use on Applied Biosystems™ 7500 FastDx RT-PCR, QuantStudioDx RT-PCR, Cepheid™ SmartCycler™ II

  • RT-PCR
  • One-step reagent setup
  • Results in <70 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Long shelf-life
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls

Specifications

Certifications/Compliance FDA approved*
Content And Storage 2°C to 8°C
Control Sets Included
Description Qualitative detection and identification of respiratory syncytial virus (RSV) and human metapneumovirus (hMPV) ribonucleic acid (RNA) extracted from nasal and nasopharyngeal swab specimens from patients with signs and symptoms of respiratory infection
CE Marker Yes
Clia Complexity High
Detectable Analytes Respiratory Syncytial Virus RNA, Human Metapneumovirus (hMPV) RNA
Sample Type Throat swab specimens obtained from patients symptomatic of acute pharyngitis
Sensitivity Group A β-hemolytic Streptococcus 96%, Pyogenic Group C and G β-hemolytic Streptococcus 95%, Positive Percent Agreement: hMPV: 98.0%
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) FDA approved for use on Applied Biosystems™ 7500 Fast Dx Real-Time PCR Instrument, QuantStudioTM Dx Real-Time PCR Instrument and Cepheid™ SmartCycler™ II
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Specificity Group A β-hemolytic Streptococcus 98%, Pyogenic Group C and G β-hemolytic Streptococcus 1.8x104 CFU/mL, Negative Percent Agreement: hMPV: 98.9%
Test Time <70 min. after extraction
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.