Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra SARS-CoV-2 Molecular Assay
SDP

Catalog No. 23044011 Shop All QuidelOrtho Products
Click to view available options
Format:
Control
Microfuge
Microwell
This item is not returnable. View return policy
This item is not returnable. View return policy

For in vitro qualitative detection of human coronavirus SARS-CoV-2 from viral RNA extracted from nasal, nasopharyngeal (NP) or oropharyngeal (OP) swab specimens from patients with signs and symptoms of COVID-19, Assay targets non-structural Polyprotein (pp1ab) of SARS-CoV-2 virus

  • RT-PCR
  • One-step reagent setup
  • Results in <75 minutes after extraction
  • No freezer necessary
  • Refrigration storage
  • Flexible testing format
  • Includes 12 x 8 master mix, rehydration solution and liquid process controls

Specifications

Certifications/Compliance Emergency Use Authorization (EUA)
Content And Storage 2°C to 8°C
Control Sets Included
CE Marker Yes
Clia Complexity High
Detectable Analytes SARS-CoV-2
DoA Calibrators No
Format Control
Sample Type Nasal, Nasopharyngeal, Oropharyngeal Swab
Sensitivity PPA – 92/92 = 100% (95% CI: 95.99 to 100%)
Type Molecular Testing
For Use With (Application) RT-PCR
For Use With (Equipment) Emergency Use Authorization (EUA) for use on bioMerieux NucliSENS™ easyMAG™ system or EMAG system, followed by RT-PCR on one of six thermocyclers: Applied Biosystems™ 7500 Fast Dx, Applied Biosystems 7500 Standard, QuantStudioTM Real-Time PCR Instrument, Roche LightCycler™ 480, BioRad CFX96 Touch, or Qiagen Rotor-Gene Q
Includes Tests and controls
Packaging Type Kit
Quantity 96 Tests/Kit
Specificity NPA – 92/92 = 100% (95% CI: 95.99 to 100%)
Test Time <75 min. after extraction
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.