Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYRIC Monoclonal Antibody (C6-C10)
GREENER_CHOICE

Catalog No. PIMA536100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536100 Supplier Invitrogen™ Supplier No. MA536100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LYRIC Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYRIC
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C6-C10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MTDH
Gene Accession No. Q80WJ7, Q86UE4, Q9Z1W6
Gene Alias 2610103J23Rik; 3D3; 3D3/Lyric; AEG1; AEG-1; astrocyte elevated gene 1; astrocyte elevated gene-1 protein; AV353288; D8Bwg1112e; LYRIC; LYRIC/ MTDH; LYRIC/3D3; lysine-rich CEACAM1 co-isolated protein; Metadherin; metastasis adhesion protein; MTDH; Protein LYRIC
Gene Symbols MTDH
Host Species Mouse
Immunogen Recombinant protein within Human LYRIC aa 6-233.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170910, 67154, 92140
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.