Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566281 Supplier Invitrogen™ Supplier No. PA566281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTTATRQEVLG LYRSIFRLAR KWQATSGQME DTIKEKQYIL NEARTLFRKN KNLTDTDLIK Highest antigen sequence identity to the following orthologs - mouse 87%, rat 92%.

LYRM1 was one of many genes identified through a suppression subtractive hybridization comparing their expression in omental adipose tissue of obese patients compared to non-obese individuals. Further study confirms that both mRNA and protein levels of LYRM1 are higher in obese individuals. LYRM1 is widely expressed, with highest levels occurring in adipose and liver tissues. It is thought that LYRM1 promotes preadipocyte proliferation and can inhibit apoptosis of preadipocytes. Ectopic LYRM1 expression did not significantly affect adipogenesis, suggesting that LYRM1 can influence adipose tissue homeostasis by modulating the size of the preadipocyte pool.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYRM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lyrm1
Gene Accession No. O43325
Gene Alias 1110065L10Rik; 2310004B22Rik; 4930404J24Rik; A211C6.1; LYR motif containing 1; LYR motif-containing protein 1; LYRM1; RGD1563498
Gene Symbols Lyrm1
Host Species Rabbit
Immunogen Recombinant Human LYRM1. Recombinant protein control fragment (Product #RP-106966).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.