Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYRM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564141 Supplier Invitrogen™ Supplier No. PA564141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FIRRQQILLL YRRILQTIWQ VPNDSDCKYL KDWAREEFRR NKTATEEDTI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

The Lyr motif found in the LYR-motif containing protein family is similar to that found in the sacchromyces cerevisiae protein ISD11, an iron-sulfur protein in the mitochondria that is thought to play a role in iron homeostasis. No known function has been assigned to LYRM2, although LYRM1 is thought to be involved in preadipocyte progression and LYRM3 has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYRM2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LYRM2
Gene Accession No. Q9NU23
Gene Alias 2610208E05Rik; DJ122O8.2; LYR motif containing 2; LYR motif-containing protein 2; Lyrm2; novel complex 1 (LYR family) protein
Gene Symbols LYRM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYRM2. Recombinant protein control fragment (Product #RP-103536).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.